Iproniazid ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

I0893-1G I0893-200MG
TCI0893-1GEA 167 USD
TCI0893-1G TCI0893-200MG
Iproniazid ≥96.0% (by HPLC, titration analysis)
Iproniazid
Formula: C₉H₁₃N₃O
MDL Number: MFCD00153819
CAS Number: 54-92-2
EINECS: 200-218-8
Merck Index: 14,05078

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 109.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR