Ethylene Glycol Di(m-tolyl) ether ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,1'-[1,2-Ethanediylbis(oxy)]bis(3-methylbenzene), 1,2-Bis(3-methylphenoxy)ethane

E1335-25G E1335-5G
TCE1335-25GEA 32 USD
TCE1335-25G TCE1335-5G
Ethylene Glycol Di(m-tolyl) ether ≥97.0% (by GC)
Ethylene Glycol Di(m-tolyl) ether
Formula: C₁₆H₁₈O₂
MW: 242.32 g/mol
MDL Number: MFCD00025952
CAS Number: 54914-85-1
EINECS: 402-730-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 96.0 to 100.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR