10-Phenyl-9(10H)-acridone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 10-Phenylacridin-9(10H)-one

P2537-1G P2537-5G
TCP2537-1GEA 96 USD
TCP2537-1G TCP2537-5G
10-Phenyl-9(10H)-acridone ≥98.0% (by HPLC)
10-Phenyl-9(10H)-acridone
Formula: C₁₉H₁₃NO
MDL Number: MFCD01463673
CAS Number: 5472-23-1

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR