N,N-Dimethyl-2-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D5605-1G D5605-5G
TCD5605-1GEA 123 USD
TCD5605-1G TCD5605-5G
N,N-Dimethyl-2-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
N,N-Dimethyl-2-(trifluoromethyl)aniline
Formula: C₉H₁₀F₃N
MW: 189.18 g/mol
Boiling Pt: 170 °C
MDL Number: MFCD01862096
CAS Number: 54672-14-9

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR