2-Phenylethyl n-Octanoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: n-Octanoic acid 2-Phenylethyl ester, Phenethyl n-Octanoate, n-Octanoic acid Phenethyl ester

P2741-100G P2741-25G
TCP2741-100GEA 114 USD
TCP2741-100G TCP2741-25G
2-Phenylethyl n-Octanoate ≥98.0% (by GC)
2-Phenylethyl n-Octanoate
Formula: C₁₆H₂₄O₂
MW: 248.37 g/mol
Boiling Pt: 296 °C
MDL Number: MFCD00209536
CAS Number: 5457-70-5
EINECS: 226-718-6

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR