Etretinate ≥98.0% (by HPLC)

Supplier: TCI America
Danger

Synonyms: Ethyl all-trans-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoate, all-trans-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoic acid Ethyl ester, Acitretin ethyl ester

E1293-250MG E1293-50MG
TCE1293-250MGEA 598 USD
TCE1293-250MG TCE1293-50MG
Etretinate ≥98.0% (by HPLC)
Etretinate
Formula: C₂₃H₃₀O₃
MW: 354.49 g/mol
MDL Number: MFCD00866624
CAS Number: 54350-48-0
EINECS: 259-119-3
UN: 3077
ADR: 9,III
Merck Index: 14,03892

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR