Isopentylamine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 3-Methylbutan-1-amine hydrochloride, Isopentylammonium chloride

I0083-1G I0083-5G
TCI0083-1GEA 54 USD
TCI0083-1G TCI0083-5G
Isopentylamine hydrochloride ≥98.0% (by titrimetric analysis)
Isopentylamine hydrochloride
Formula: C₅H₁₃N·HCl
MW: 123.62 g/mol
MDL Number: MFCD00050684
CAS Number: 541-23-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR