3-Methoxy-N,N-dimethylpropanamide ≥98.0% (by GC)

Supplier: TCI America
Warning

M3253-100G M3253-25G
TCM3253-100GEA 183 USD
TCM3253-100G TCM3253-25G
3-Methoxy-N,N-dimethylpropanamide ≥98.0% (by GC)
3-Methoxy-N,N-dimethylpropanamide
Formula: C₆H₁₃NO₂
Boiling Pt: 58 °C
MDL Number: MFCD18970488
CAS Number: 53185-52-7
EINECS: 258-420-7

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR