N,N,N-Trimethyl-1-adamantylammonium hydroxide 25% in water

Supplier: TCI America
Danger

T3821-100G T3821-25G
TCT3821-100GEA 262 USD
TCT3821-100G TCT3821-25G
N,N,N-Trimethyl-1-adamantylammonium hydroxide 25% in water
N,N,N-Trimethyl-1-adamantylammonium hydroxide
Formula: C₁₃H₂₅NO
MDL Number: MFCD27920795
CAS Number: 53075-09-5
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Concentration (Nonaqueous Titration) 25.0 to 28.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR