Polypropylene Glycol Diacrylate n=∼12 stabilised

Supplier: TCI America
Warning

Synonyms: PPG Diacrylate, PPGDA

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2759-25G P2759-500G
TCP2759-25GEA 28 USD
TCP2759-25G TCP2759-500G
Polypropylene Glycol Diacrylate n=∼12 stabilised
Polypropylene Glycol Diacrylate
Storage Temperature: Refrigerator
MDL Number: MFCD00081912
CAS Number: 52496-08-9

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
The average value of n 11.0 to 13.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR