Mesityleneruthenium(II) chloride Dimer ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Dichloro(mesitylene)ruthenium(II) Dimer

D5524-1G D5524-250MG
TCD5524-1GEA 329 USD
TCD5524-1G TCD5524-250MG
Mesityleneruthenium(II) chloride Dimer ≥97.0% (by titrimetric analysis)
Mesityleneruthenium(II) chloride Dimer
Formula: C₁₈H₂₄Cl₄Ru₂
MDL Number: MFCD11656079
CAS Number: 52462-31-4

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR