Ethylene Glycol bis(2-aminophenyl) ether ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1,2-Bis(2-aminophenoxy)ethane, 2,2'-[Ethane-1,2-diylbis(oxy)]dianiline, 1,2-Bis(o-aminophenoxy)ethane

E1365-1G E1365-5G
TCE1365-1GEA 51 USD
TCE1365-1G TCE1365-5G
Ethylene Glycol bis(2-aminophenyl) ether ≥98.0% (by HPLC, titration analysis)
Ethylene Glycol bis(2-aminophenyl) ether
Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
Boiling Pt: 225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00087648
CAS Number: 52411-34-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR