N,N-Dimethyl-1,3-propanediamine dihydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: N1,N1-Dimethylpropane-1,3-diamine dihydrochloride, N,N-Dimethyl-1,3-propanediammonium Dichloride, 3-(Dimethylamino)propylamine dihydrochloride

D5617-1G D5617-5G
TCD5617-1GEA 34 USD
TCD5617-1G TCD5617-5G
N,N-Dimethyl-1,3-propanediamine dihydrochloride ≥98.0% (by titrimetric analysis)
N,N-Dimethyl-1,3-propanediamine dihydrochloride
Formula: C₅H₁₄N₂·₂HCl
MW: 175.10 g/mol
MDL Number: MFCD32062892
CAS Number: 52198-63-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 187.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR