Isononyl acrylate (mixture of branched chain isomers) stabilized

Supplier: TCI America
Warning

Synonyms: Acrylic acid Isononyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

I1071-100ML I1071-500ML
TCI1071-100MLEA 31 USD
TCI1071-100ML TCI1071-500ML
Isononyl acrylate (mixture of branched chain isomers) stabilized
Isononyl acrylate (mixture of branched chain isomers)
Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
CAS Number: 51952-49-9
EINECS: 257-544-9

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Average alkylchain length 8.5 to 9.4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR