Dibenzo[b,d]furan-4-amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Dibenzofuranamine, 4-Aminodibenzofuran

D5520-5G D5520-1G
TCD5520-5GEA 372 USD
TCD5520-5G TCD5520-1G
Dibenzo[b,d]furan-4-amine ≥98.0% (by GC, titration analysis)
Dibenzo[b,d]furan-4-amine
Formula: C₁₂H₉NO
Melting Pt: 85 °C
MDL Number: MFCD11110988
CAS Number: 50548-43-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR