(Triphenylmethyl)thionyl Imide ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Tritylthionyl Imide, TrNSO

T3773-1G T3773-5G
TCT3773-1GEA 91 USD
TCT3773-1G TCT3773-5G
(Triphenylmethyl)thionyl Imide ≥97.0% (by GC)
(Triphenylmethyl)thionyl Imide
Formula: C₁₉H₁₅NOS
MDL Number: MFCD32062442
CAS Number: 503596-47-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR