2-(2-Ethylhexyl)thiophene ≥98.0% (by GC)

Supplier: TCI America

E1294-1ML E1294-5ML
TCE1294-1MLEA 72 USD
TCE1294-1ML TCE1294-5ML
2-(2-Ethylhexyl)thiophene ≥98.0% (by GC)
2-(2-Ethylhexyl)thiophene
Formula: C₁₂H₂₀S
Storage Temperature: Refrigerator
MDL Number: MFCD26743668
CAS Number: 4891-44-5

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR