N,N,N',N'-Tetrakis[4-(diisobutylamino)phenyl]-1,4-phenylenediamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N1,N1'-(1,4-Phenylene)bis(N1-(4-(diisobutylamino)phenyl)-N4,N4-diisobutylbenzene-1,4-diamine), N1,N1'-(1,4-Phenylene)bis[N1-[4-(diisobutylamino)phenyl]-N4,N4-diisobutylbenzene-1,4-diamine]

T3770-1G T3770-5G
TCT3770-1GEA 94 USD
TCT3770-1G TCT3770-5G
N,N,N',N'-Tetrakis[4-(diisobutylamino)phenyl]-1,4-phenylenediamine ≥98.0% (by titrimetric analysis)
N,N,N',N'-Tetrakis[4-(diisobutylamino)phenyl]-1,4-phenylenediamine
Formula: C₆₂H₉₂N₆
MDL Number: MFCD22207043
CAS Number: 485831-34-3

Order Now

Specification Test Results

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 177.0 to 181.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR