9-(2-Ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 9-(2-Ethylhexyl)-9H-carbazole-2,7-diboronic acid bis(pinacol) ester

E1324-1G E1324-200MG
TCE1324-1GEA 500 USD
TCE1324-1G TCE1324-200MG
9-(2-Ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC, titration analysis)
9-(2-Ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Formula: C₃₂H₄₇B₂NO₄
MDL Number: MFCD23703117
CAS Number: 476360-83-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR