2-Fluoroethylamine hydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Aminoethyl Fluoride hydrochloride, 2-Fluoroethanamine hydrochloride, 2-Fluoroethylammonium hydrochloride

F1250-1G F1250-5G
TCF1250-1GEA 44 USD
TCF1250-1G TCF1250-5G
2-Fluoroethylamine hydrochloride ≥97.0% (by titrimetric analysis)
2-Fluoroethylamine hydrochloride
Formula: C₂H₆FN·HCl
Melting Pt: 100 °C
MDL Number: MFCD00008161
CAS Number: 460-08-2
EINECS: 207-302-3

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR