Potassium Butyltrifluoroborate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Potassium butyltrifluoroborate

P2626-1G P2626-5G
TCP2626-1GEA 39 USD
TCP2626-1G TCP2626-5G
Potassium Butyltrifluoroborate ≥98.0% (by titrimetric analysis)
Potassium Butyltrifluoroborate
Formula: C₄H₉BF₃K
MW: 164.02 g/mol
MDL Number: MFCD08276805
CAS Number: 444343-55-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Gravimetric) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR