2-Heptyl-1,3-dioxolane ≥97.0% (by GC)

Supplier: TCI America

H1715-25G H1715-5G
TCH1715-25GEA 303 USD
TCH1715-25G TCH1715-5G
2-Heptyl-1,3-dioxolane ≥97.0% (by GC)
2-Heptyl-1,3-dioxolane
Formula: C₁₀H₂₀O₂
Boiling Pt: 97 °C
MDL Number: MFCD00216992
CAS Number: 4359-57-3
EINECS: 224-438-9

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR