Hexaethylene Glycol Di-p-toluenesulfonate ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

H1681-1G H1681-5G
TCH1681-1GEA 60 USD
TCH1681-1G TCH1681-5G
Hexaethylene Glycol Di-p-toluenesulfonate ≥95.0% (by HPLC, titration analysis)
Hexaethylene Glycol Di-p-toluenesulfonate
Formula: C₂₆H₃₈O₁₁S₂
MDL Number: MFCD00319275
CAS Number: 42749-27-9

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 95.0 area%
Purity (Ester Value) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR