Isoorientin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Homoorientin, Luteolin 6-C-beta-D-Glucoside

I1087-10MG I1087-50MG
TCI1087-10MGEA 189 USD
TCI1087-10MG TCI1087-50MG
Isoorientin ≥98.0% (by HPLC)
Isoorientin
Formula: C₂₁H₂₀O₁₁
MW: 448.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017433
CAS Number: 4261-42-1

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Carbon) 53.40 to 57.40 %
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR