1-Ethyl-4-isopropylbenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Isopropylethylbenzene

E1406-1ML E1406-5ML
TCE1406-1MLEA 121 USD
TCE1406-1ML TCE1406-5ML
1-Ethyl-4-isopropylbenzene ≥98.0% (by GC)
1-Ethyl-4-isopropylbenzene
Formula: C₁₁H₁₆
MW: 148.25 g/mol
Melting Pt: 197 °C
MDL Number: MFCD00061006
CAS Number: 4218-48-8

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR