N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N1,N1'-(1,4-Phenylene)bis(N4,N4-dibutyl-N1-(4-(dibutylamino)phenyl)benzene-1,4-diamine), N1,N1'-(1,4-Phenylene)bis[N4,N4-dibutyl-N1-[4-(dibutylamino)phenyl]benzene-1,4-diamine]

T3876-1G T3876-5G
TCT3876-1GEA 98 USD
TCT3876-1G TCT3876-5G
N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine ≥98.0% (by HPLC)
N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine
Formula: C₆₂H₉₂N₆
MW: 921.46 g/mol
MDL Number: MFCD09832942
CAS Number: 4182-80-3
EINECS: 224-057-8

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 103.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR