Diethyl 2,4-Pyridinedicarboxylate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Diethyl pyridine-2,4-dicarboxylate, 2,4-Pyridinedicarboxylic acid Diethyl ester

D5518-1G D5518-200MG
TCD5518-1GEA 170 USD
TCD5518-1G TCD5518-200MG
Diethyl 2,4-Pyridinedicarboxylate ≥97.0% (by GC)
Diethyl 2,4-Pyridinedicarboxylate
Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Boiling Pt: 180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00152168
CAS Number: 41438-38-4

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR