4'-Hydroxyflavone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(4-Hydroxyphenyl)-4H-chromen-4-one, 2-(4-Hydroxyphenyl)-4H-1-benzopyran-4-one

H1638-1G H1638-5G
TCH1638-1GEA 123 USD
TCH1638-1G TCH1638-5G
4'-Hydroxyflavone ≥98.0% (by GC, titration analysis)
4'-Hydroxyflavone
Formula: C₁₅H₁₀O₃
MW: 238.24 g/mol
MDL Number: MFCD00017687
CAS Number: 4143-63-9

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR