4',5'-Dimethoxy-2'-nitroacetophenone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(4,5-Dimethoxy-2-nitrophenyl)ethanone

D5477-1G D5477-5G
TCD5477-1GEA 51 USD
TCD5477-1G TCD5477-5G
4',5'-Dimethoxy-2'-nitroacetophenone ≥98.0% (by GC)
4',5'-Dimethoxy-2'-nitroacetophenone
Formula: C₁₀H₁₁NO₅
MW: 225.20 g/mol
MDL Number: MFCD00130048
CAS Number: 4101-32-0

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR