1-Ethyl-2-methylindole ≥98.0% (by GC)

Supplier: TCI America

E1376-25G E1376-5G
TCE1376-25GEA 230 USD
TCE1376-25G TCE1376-5G
1-Ethyl-2-methylindole ≥98.0% (by GC)
1-Ethyl-2-methylindole
Formula: C₁₁H₁₃N
Boiling Pt: 270 °C
MDL Number: MFCD01632168
CAS Number: 40876-94-6
EINECS: 255-121-3

Order Now

Specification Test Results

Appearance White to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR