Ethyl 2,3-Dicyanopropionate ≥98.0% (by GC)

Supplier: TCI America
Warning

E1366-25G E1366-5G
TCE1366-25GEA 138 USD
TCE1366-25G TCE1366-5G
Ethyl 2,3-Dicyanopropionate ≥98.0% (by GC)
Ethyl 2,3-Dicyanopropionate
Formula: C₇H₈N₂O₂
Boiling Pt: 140 °C
MDL Number: MFCD08061844
CAS Number: 40497-11-8

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR