(Trifluoromethyl)cyclohexane ≥98.0% (by GC)

Supplier: TCI America
Danger

T3628-25G T3628-5G
TCT3628-25GEA 125 USD
TCT3628-25G TCT3628-5G
(Trifluoromethyl)cyclohexane ≥98.0% (by GC)
(Trifluoromethyl)cyclohexane
Formula: C₇H₁₁F₃
Boiling Pt: 107 °C
MDL Number: MFCD03412289
CAS Number: 401-75-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR