1-Isopropoxy-2-propanol ≥98.0% (by GC)

Supplier: TCI America
Warning

I1084-1G I1084-5G
TCI1084-1GEA 115 USD
TCI1084-1G TCI1084-5G
1-Isopropoxy-2-propanol ≥98.0% (by GC)
1-Isopropoxy-2-propanol
Formula: C₆H₁₄O₂
Boiling Pt: 137 °C
MDL Number: MFCD06252288
CAS Number: 3944-36-3
EINECS: 223-533-2
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR