2,7-Dibromodibenzo[b,e][1,4]dioxin ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,7-Dibromodibenzo[b,e][1,4]dioxine

D5528-1G D5528-200MG
TCD5528-1GEA 353 USD
TCD5528-1G TCD5528-200MG
2,7-Dibromodibenzo[b,e][1,4]dioxin ≥98.0% (by GC)
2,7-Dibromodibenzo[b,e][1,4]dioxin
Formula: C₁₂H₆Br₂O₂
MW: 341.99 g/mol
CAS Number: 39073-07-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 194.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR