3,4-Diethoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D5478-1G D5478-5G
TCD5478-1GEA 110 USD
TCD5478-1G TCD5478-5G
3,4-Diethoxyaniline ≥98.0% (by GC, titration analysis)
3,4-Diethoxyaniline
Formula: C₁₀H₁₅NO₂
MDL Number: MFCD00026782
CAS Number: 39052-12-5

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR