2,4,6-Trimethyl-2,4,6-trivinylcyclotrisiloxane ≥95.0% (by GC)

Supplier: TCI America

T3745-5G T3745-25G
TCT3745-5GEA 34 USD
TCT3745-5G TCT3745-25G
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisiloxane ≥95.0% (by GC)
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisiloxane
Formula: C₉H₁₈O₃Si₃
Boiling Pt: 80 °C
MDL Number: MFCD00023111
CAS Number: 3901-77-7
EINECS: 223-458-5

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 0.9690 to 0.9790
Refractive index n20/D 1.4240 to 1.4280


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR