(3-Ethyloxetan-3-yl)methyl methacrylate ≥98.0% (by GC) stabilised

Supplier: TCI America

Synonyms: Methacrylic acid (3-Ethyloxetan-3-yl)methyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

E1379-1G E1379-5G
TCE1379-1GEA 104 USD
TCE1379-1G TCE1379-5G
(3-Ethyloxetan-3-yl)methyl methacrylate ≥98.0% (by GC) stabilised
(3-Ethyloxetan-3-yl)methyl methacrylate
Formula: C₁₀H₁₆O₃
MW: 184.24 g/mol
MDL Number: MFCD16039335
CAS Number: 37674-57-0

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR