2-Dibenzofuranamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D5494-1G D5494-5G
TCD5494-1GEA 83 USD
TCD5494-1G TCD5494-5G
2-Dibenzofuranamine ≥98.0% (by GC, titration analysis)
2-Dibenzofuranamine
Formula: C₁₂H₉NO
MDL Number: MFCD00033416
CAS Number: 3693-22-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR