Tris[2-(3-mercaptopropionyloxy)ethyl] Isocyanurate ≥70% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Isocyanuric acid tris[2-(3-mercaptopropionyloxy)ethyl] ester

T3484-25G T3484-500G
TCT3484-25GEA 46 USD
TCT3484-25G TCT3484-500G
Tris[2-(3-mercaptopropionyloxy)ethyl] Isocyanurate ≥70% (by HPLC)
Tris[2-(3-mercaptopropionyloxy)ethyl] Isocyanurate
Formula: C₁₈H₂₇N₃O₉S₃
MW: 525.61 g/mol
MDL Number: MFCD04038141
CAS Number: 36196-44-8
EINECS: 252-907-8

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 70.0 area%
Purity (Iodometric Titration) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR