2-Methyl-N-(m-tolyl)aniline ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Methyl-N-(3-methylphenyl)benzenamine, 2,3'-Dimethyldiphenylamine, 2-Methyl-N-(3-methylphenyl)aniline

M3064-1G M3064-5G
TCM3064-1GEA 149 USD
TCM3064-1G TCM3064-5G
2-Methyl-N-(m-tolyl)aniline ≥98.0% (by GC)
2-Methyl-N-(m-tolyl)aniline
Formula: C₁₄H₁₅N
Boiling Pt: 187 °C
CAS Number: 34801-11-1

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR