3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl ≥99.0% (by HPLC, total nitrogen), purified by sublimation

Supplier: TCI America
Danger

Synonyms: mCBP

D5594-1G D5594-200MG
TCD5594-1GEA 335 USD
TCD5594-1G TCD5594-200MG
3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl ≥99.0% (by HPLC, total nitrogen), purified by sublimation
3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl
Formula: C₃₆H₂₄N₂
MW: 484.60 g/mol
MDL Number: MFCD28138084
CAS Number: 342638-54-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR