Dodecylamine Hydroiodide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

Synonyms: Dodecylammonium iodide, Dodecan-1-amine Hydroiodide, Laurylamine Hydroiodide, 1-Aminododecane Hydroiodide

D5538-1G D5538-5G
TCD5538-1GEA 54 USD
TCD5538-1G TCD5538-5G
Dodecylamine Hydroiodide ≥98.0% (by total nitrogen and titration analysis)
Dodecylamine Hydroiodide
Formula: C₁₂H₂₇N·HI
MW: 313.27 g/mol
CAS Number: 34099-97-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR