DL-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

L0228-25G L0228-5G
TCL0228-25GEA 222 USD
TCL0228-25G TCL0228-5G
DL-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen basis)
DL-Lysine methyl ester dihydrochloride
Formula: C₇H₁₆N₂O₂·₂HCl
MDL Number: MFCD03547976
CAS Number: 34015-48-0
EINECS: 251-794-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -1.0 to +1.0 deg (C=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR