Tebuthiuron ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-[5-(tert-Butyl)-1,3,4-thiadiazol-2-yl]-1,3-dimethylurea

T3647-25G T3647-5G
TCT3647-25GEA 664 USD
TCT3647-25G TCT3647-5G
Tebuthiuron ≥98.0% (by GC)
Tebuthiuron
Formula: C₉H₁₆N₄OS
MW: 228.31 g/mol
MDL Number: MFCD00078732
CAS Number: 34014-18-1
EINECS: 251-793-7
UN: 3077
ADR: 9,III
Merck Index: 14,09095

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 161.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR