6-Undecanamine ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

U0094-1ML U0094-5ML
TCU0094-1MLEA 171 USD
TCU0094-1ML TCU0094-5ML
6-Undecanamine ≥97.0% (by titrimetric analysis)
6-Undecanamine
Formula: C₁₁H₂₅N
Boiling Pt: 158 °C
MDL Number: MFCD00047924
CAS Number: 33788-00-0
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR