N,N-Dimethyl-3-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N,N-Dimethyl-3-trifluoromethylaniline

D5606-1G D5606-5G
TCD5606-1GEA 119 USD
TCD5606-1G TCD5606-5G
N,N-Dimethyl-3-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
N,N-Dimethyl-3-(trifluoromethyl)aniline
Formula: C₉H₁₀F₃N
MW: 189.18 g/mol
Boiling Pt: 228 °C
MDL Number: MFCD01632487
CAS Number: 329-00-0

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR