9-Ethynylphenanthrene ≥98.0% (by GC)

Supplier: TCI America

E1301-1G E1301-5G
TCE1301-1GEA 119 USD
TCE1301-1G TCE1301-5G
9-Ethynylphenanthrene ≥98.0% (by GC)
9-Ethynylphenanthrene
Formula: C₁₆H₁₀
Boiling Pt: 377 °C
MDL Number: MFCD05664210
CAS Number: 32870-98-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR