1-Ethylcyclopentyl acrylate ≥95.0% (by GC) stabilized

Supplier: TCI America


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

E1405-1G E1405-5G
TCE1405-1GEA 103 USD
TCE1405-1G TCE1405-5G
1-Ethylcyclopentyl acrylate ≥95.0% (by GC) stabilized
1-Ethylcyclopentyl acrylate
Formula: C₁₀H₁₆O₂
Storage Temperature: Refrigerator
MDL Number: MFCD31716062
CAS Number: 326925-69-3

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR