trans-2-Dodecenoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: (E)-Dodec-2-enoic acid

D5660-1G D5660-250MG
TCD5660-1GEA 498 USD
TCD5660-1G TCD5660-250MG
trans-2-Dodecenoic acid ≥98.0% (by GC, titration analysis)
trans-2-Dodecenoic acid
Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
Boiling Pt: 146 °C
Melting Pt: 23 °C
MDL Number: MFCD00045906
CAS Number: 32466-54-9

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR