Heptadecan-9-amine ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 1-Octylnonylamine, 9-Aminoheptadecane

H1640-1ML H1640-5ML
TCH1640-1MLEA 193 USD
TCH1640-1ML TCH1640-5ML
Heptadecan-9-amine ≥96.0% (by GC, titration analysis)
Heptadecan-9-amine
Formula: C₁₇H₃₇N
MW: 255.49 g/mol
Boiling Pt: 120 °C
MDL Number: MFCD30742905
CAS Number: 3241-20-1
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR